3-[(3-Chlorobenzyl)sulfanyl]-1H-1,2,4-triazol-5-ylamine ≥95%

Supplier: Matrix Scientific

045261-500MG 045261-1G 045261-5G
101869-984EA 181.54 USD
101869-984 101869-986 101869-988
3-[(3-Chlorobenzyl)sulfanyl]-1H-1,2,4-triazol-5-ylamine ≥95%
3-[(3-Chlorobenzyl)sulfanyl]-1H-1,2,4-triazol-5-ylamine

Hazard: Irritant

Formula: C₉H₉ClN₄S
Melting Pt: 116…118 °C
MDL Number: MFCD00215050
CAS Number: 344262-76-6

Order Now

Specification Test Results

Melting point 116-118°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR