3-[(2-Chlorobenzyl)sulfanyl]-1H-1,2,4-triazol-5-ylamine ≥95%

Supplier: Matrix Scientific

045260-500MG 045260-1G 045260-5G
101869-978EA 181.54 USD
101869-978 101869-980 101869-982
3-[(2-Chlorobenzyl)sulfanyl]-1H-1,2,4-triazol-5-ylamine ≥95%
3-[(2-Chlorobenzyl)sulfanyl]-1H-1,2,4-triazol-5-ylamine

Hazard: Irritant

Formula: C₉H₉ClN₄S
Melting Pt: 142…144 °C
MDL Number: MFCD00215049
CAS Number: 341944-06-7

Order Now

Specification Test Results

Melting point 142-144°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR