4-Acetyl-1-methyl-1H-pyrrole-2-carboxylic acid ≥95%
Supplier: Matrix Scientific
045258-500MG
045258-1G
045258-5G
101869-966EA
907.69
USD
101869-966
101869-968
101869-970
4-Acetyl-1-methyl-1H-pyrrole-2-carboxylic acid ≥95%
4-Acetyl-1-methyl-1H-pyrrole-2-carboxylic acid
Hazard: Irritant
Formula:
C₈H₉NO₃ Melting Pt: 187…189 °C |
MDL Number:
MFCD00665069 CAS Number: 339011-93-7 |
Specification Test Results
Melting point | 187-189°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...