2,3-Dibromo-3-phenylpropiophenone ≥95%

Supplier: Matrix Scientific

045255-500MG 045255-1G
101869-950EA 181.54 USD
101869-950 101869-952
2,3-Dibromo-3-phenylpropiophenone ≥95%
2,3-Dibromo-3-phenylpropiophenone

Hazard: Irritant

Formula: C₁₅H₁₂Br₂O
Melting Pt: 156-158°C
MDL Number: MFCD00017861
CAS Number: 611-91-6

Order Now

Specification Test Results

Melting point 156-158°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR