4-[(4-Fluorophenyl)sulfanyl]aniline ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(4-Fluorophenylthio)aniline

045254-500MG 045254-1G 045254-5G
101869-944EA 747.69 USD
101869-944 101869-946 101869-948
4-[(4-Fluorophenyl)sulfanyl]aniline ≥95%
4-[(4-Fluorophenyl)sulfanyl]aniline

Hazard: Irritant

Formula: C₁₂H₁₀FNS
MW: 219.28 g/mol
Melting Pt: 60…62 °C
MDL Number: MFCD00172068
CAS Number: 24900-69-4

Order Now

Specification Test Results

Melting point 60-62°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR