1-Fluoro-4-[(4-nitrophenyl)sulfanyl]benzene ≥95%

Supplier: Matrix Scientific

Synonyms: (4-Fluorophenyl)(4-nitrophenyl)sulphane, 4-[(4-Fluorophenyl)thio]nitrobenzene

045253-500MG 045253-1G 045253-5G
101869-938EA 181.54 USD
101869-938 101869-940 101869-942
1-Fluoro-4-[(4-nitrophenyl)sulfanyl]benzene ≥95%
1-Fluoro-4-[(4-nitrophenyl)sulfanyl]benzene

Hazard: Irritant

Formula: C₁₂H₈FNO₂S
Melting Pt: 92…95 °C
MDL Number: MFCD04117815
CAS Number: 2438-85-9

Order Now

Specification Test Results

Melting point 92-95°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR