Methyl-3-amino-1-cyano-4-fluoro-1-phenyl-1H-indene-2-carboxylate ≥95%

Supplier: Matrix Scientific

045250-500MG 045250-1G 045250-5G
101869-930EA 556.92 USD
101869-930 101869-932 101869-934
Methyl-3-amino-1-cyano-4-fluoro-1-phenyl-1H-indene-2-carboxylate ≥95%
Methyl-3-amino-1-cyano-4-fluoro-1-phenyl-1H-indene-2-carboxylate

Hazard: Irritant

Formula: C₁₈H₁₃FN₂O₂
MW: 308.31 g/mol
Melting Pt: 154…156 °C
MDL Number: MFCD00232259
CAS Number: 339010-38-7

Order Now

Specification Test Results

Melting point 154-156°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR