Ethyl-2-(2-amino-1,3-thiazol-4-yl)benzenecarboxylate ≥95%

Supplier: Matrix Scientific

045249-500MG 045249-1G 045249-5G
101869-924EA 569.23 USD
101869-924 101869-926 101869-928
Ethyl-2-(2-amino-1,3-thiazol-4-yl)benzenecarboxylate ≥95%
Ethyl-2-(2-amino-1,3-thiazol-4-yl)benzenecarboxylate

Hazard: Irritant

Formula: C₁₂H₁₂N₂O₂S
Melting Pt: 135…137 °C
MDL Number: MFCD00142010
CAS Number: 339010-09-2

Order Now

Specification Test Results

Melting point 135-137°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR