3-[(4-Chlorobenzyl)oxy]-2-thiophenecarboxylic acid ≥95%

Supplier: Matrix Scientific

045247-500MG 045247-1G 045247-5G
101869-912EA 375.38 USD
101869-912 101869-914 101869-916
3-[(4-Chlorobenzyl)oxy]-2-thiophenecarboxylic acid ≥95%
3-[(4-Chlorobenzyl)oxy]-2-thiophenecarboxylic acid

Hazard: Irritant

Formula: C₁₂H₉ClO₃S
Melting Pt: 158…159 °C
MDL Number: MFCD00173490
CAS Number: 339009-58-4

Order Now

Specification Test Results

Melting point 158-159°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR