2-(2,4-Dichlorophenyl)-3-(dimethylamino)acrylonitrile ≥95%

Supplier: Matrix Scientific

045246-500MG 045246-1G 045246-5G
101869-906EA 181.54 USD
101869-906 101869-908 101869-910
2-(2,4-Dichlorophenyl)-3-(dimethylamino)acrylonitrile ≥95%
2-(2,4-Dichlorophenyl)-3-(dimethylamino)acrylonitrile

Hazard: Irritant

Formula: C₁₁H₁₀Cl₂N₂
MW: 241.12 g/mol
Melting Pt: 66…69 °C
MDL Number: MFCD27920579
CAS Number: 339009-45-9

Order Now

Specification Test Results

Melting point 66-69°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR