3-[(4-Chlorobenzyl)oxy]-2-thiophenecarbohydrazide ≥95%

Supplier: Matrix Scientific

045245-500MG 045245-1G 045245-5G
101869-900EA 375.38 USD
101869-900 101869-902 101869-904
3-[(4-Chlorobenzyl)oxy]-2-thiophenecarbohydrazide ≥95%
3-[(4-Chlorobenzyl)oxy]-2-thiophenecarbohydrazide

Hazard: Irritant

Formula: C₁₂H₁₁ClN₂O₂S
Melting Pt: 141…143 °C
MDL Number: MFCD00173482
CAS Number: 339009-44-8

Order Now

Specification Test Results

Melting point 141-143°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR