Methyl-3-[(4-chlorobenzyl)oxy]-2-thiophenecarboxylate ≥95%

Supplier: Matrix Scientific

045243-500MG 045243-1G 045243-5G
101869-888EA 181.54 USD
101869-888 101869-890 101869-892
Methyl-3-[(4-chlorobenzyl)oxy]-2-thiophenecarboxylate ≥95%
Methyl-3-[(4-chlorobenzyl)oxy]-2-thiophenecarboxylate

Hazard: Irritant

Formula: C₁₃H₁₁ClO₃S
Melting Pt: 104…106 °C
MDL Number: MFCD00173481
CAS Number: 339009-41-5

Order Now

Specification Test Results

Melting point 104-106°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR