Ethyl-3-[(4-methylbenzoyl)amino]-2-oxo-6-phenyl-2H-pyran-5-carboxylate ≥95%

Supplier: Matrix Scientific

045241-500MG 045241-1G 045241-5G
101869-876EA 375.38 USD
101869-876 101869-878 101869-880
Ethyl-3-[(4-methylbenzoyl)amino]-2-oxo-6-phenyl-2H-pyran-5-carboxylate ≥95%
Ethyl-3-[(4-methylbenzoyl)amino]-2-oxo-6-phenyl-2H-pyran-5-carboxylate

Hazard: Irritant

Formula: C₂₂H₁₉NO₅
Melting Pt: 186…188 °C
MDL Number: MFCD00139850
CAS Number: 339009-32-4

Order Now

Specification Test Results

Melting point 186-188°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR