5-Chloro-1-(2,6-dichlorobenzyl)-2-oxo-1,2-dihydro-3-pyridine carboxylic acid ≥95%
Supplier: Matrix Scientific
045236-500MG
045236-1G
045236-5G
101869-846EA
556.92
USD
101869-846
101869-848
101869-850
5-Chloro-1-(2,6-dichlorobenzyl)-2-oxo-1,2-dihydro-3-pyridine carboxylic acid ≥95%
5-Chloro-1-(2,6-dichlorobenzyl)-2-oxo-1,2-dihydro-3-pyridine carboxylic acid
Hazard: Irritant
Formula:
C₁₃H₈Cl₃NO₃ Melting Pt: 198…200 °C |
MDL Number:
MFCD00140281 CAS Number: 339009-02-8 |
Specification Test Results
Melting point | 198-200°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...