5-Chloro-1-(3,4-dichlorobenzyl)-2-oxo-1,2-dihydro-3-pyridine carboxylic acid ≥95%

Supplier: Matrix Scientific

045234-500MG 045234-1G 045234-5G
101869-834EA 556.92 USD
101869-834 101869-836 101869-838
5-Chloro-1-(3,4-dichlorobenzyl)-2-oxo-1,2-dihydro-3-pyridine carboxylic acid ≥95%
5-Chloro-1-(3,4-dichlorobenzyl)-2-oxo-1,2-dihydro-3-pyridine carboxylic acid

Hazard: Irritant

Formula: C₁₃H₈Cl₃NO₃
Melting Pt: 208…209 °C
MDL Number: MFCD00140273
CAS Number: 339008-81-0

Order Now

Specification Test Results

Melting point 208-209°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR