3-(Benzyloxy)-2-thiophenecarbohydrazide ≥95%
Supplier: Matrix Scientific
045232-500MG
045232-1G
045232-5G
101869-822EA
375.38
USD
101869-822
101869-824
101869-826
3-(Benzyloxy)-2-thiophenecarbohydrazide ≥95%
3-(Benzyloxy)-2-thiophenecarbohydrazide
Hazard: Irritant
Formula:
C₁₂H₁₂N₂O₂S Melting Pt: 98…100 °C |
MDL Number:
MFCD00215166 CAS Number: 343375-80-4 |
Specification Test Results
Melting point | 98-100°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...