Methyl-3-[(2,4-dichlorobenzyl)oxy]-2-thiophenecarboxylate ≥95%

Supplier: Matrix Scientific

045231-500MG 045231-1G 045231-5G
101869-816EA 181.54 USD
101869-816 101869-818 101869-820
Methyl-3-[(2,4-dichlorobenzyl)oxy]-2-thiophenecarboxylate ≥95%
Methyl-3-[(2,4-dichlorobenzyl)oxy]-2-thiophenecarboxylate

Hazard: Irritant

Formula: C₁₃H₁₀Cl₂O₃S
Melting Pt: 122…123 °C
MDL Number: MFCD00202082
CAS Number: 343375-78-0

Order Now

Specification Test Results

Melting point 122-123°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR