1-[2,6-Dinitro-4-(trifluoromethyl)phenyl]piperazine ≥95%
Supplier: Matrix Scientific
045226-500MG
045226-1G
045226-5G
101869-790EA
181.54
USD
101869-790
101869-792
101869-794
1-[2,6-Dinitro-4-(trifluoromethyl)phenyl]piperazine ≥95%
1-[2,6-Dinitro-4-(trifluoromethyl)phenyl]piperazine
Hazard: Irritant
Formula:
C₁₁H₁₁F₃N₄O₄ Melting Pt: 128…130 °C |
MDL Number:
MFCD00052948 CAS Number: 36317-84-7 |
Specification Test Results
Melting point | 128-130°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...