2-[Amino(phenyl)methylene]malononitrile ≥95%
Supplier: Matrix Scientific
045225-500MG
045225-1G
045225-5G
101869-784EA
375.38
USD
101869-784
101869-786
101869-788
2-[Amino(phenyl)methylene]malononitrile ≥95%
2-[Amino(phenyl)methylene]malononitrile
Hazard: Irritant
Formula:
C₁₀H₇N₃ Melting Pt: 178…180 °C |
MDL Number:
MFCD00139205 CAS Number: 3336-65-0 |
Specification Test Results
Melting point | 178-180°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...