2-{1-Amino-2-[3-(trifluoromethyl)phenoxy]ethylidene}malononitrile ≥95%

Supplier: Matrix Scientific

045224-500MG 045224-1G 045224-5G
101869-778EA 375.38 USD
101869-778 101869-780 101869-782
2-{1-Amino-2-[3-(trifluoromethyl)phenoxy]ethylidene}malononitrile ≥95%
2-{1-Amino-2-[3-(trifluoromethyl)phenoxy]ethylidene}malononitrile

Hazard: Irritant

Formula: C₁₂H₈F₃N₃O
Melting Pt: 205…207 °C
MDL Number: MFCD00139203
CAS Number: 343375-42-8

Order Now

Specification Test Results

Melting point 205-207°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR