2-[1-Amino-2-(4-nitrophenoxy)ethylidene]malononitrile ≥95%

Supplier: Matrix Scientific

045223-500MG 045223-1G 045223-5G
101869-772EA 375.38 USD
101869-772 101869-774 101869-776
2-[1-Amino-2-(4-nitrophenoxy)ethylidene]malononitrile ≥95%
2-[1-Amino-2-(4-nitrophenoxy)ethylidene]malononitrile

Hazard: Irritant

Formula: C₁₁H₈N₄O₃
MW: 244.21 g/mol
Melting Pt: 237…239 °C
MDL Number: MFCD00139202
CAS Number: 343375-41-7

Order Now

Specification Test Results

Melting point 237-239°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR