2-[1-Amino-2-(3-chlorophenoxy)ethylidene]malononitrile ≥95%

Supplier: Matrix Scientific

045221-500MG 045221-1G 045221-5G
101869-760EA 375.38 USD
101869-760 101869-762 101869-764
2-[1-Amino-2-(3-chlorophenoxy)ethylidene]malononitrile ≥95%
2-[1-Amino-2-(3-chlorophenoxy)ethylidene]malononitrile

Hazard: Irritant

Formula: C₁₁H₈ClN₃O
MW: 233.66 g/mol
Melting Pt: 203…205 °C
MDL Number: MFCD00139201
CAS Number: 343375-39-3

Order Now

Specification Test Results

Melting point 203-205°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR