2-[1-Amino-2-(4-fluorophenoxy)ethylidene]malononitrile ≥95%

Supplier: Matrix Scientific

045220-500MG 045220-1G 045220-5G
101869-754EA 375.38 USD
101869-754 101869-756 101869-758
2-[1-Amino-2-(4-fluorophenoxy)ethylidene]malononitrile ≥95%
2-[1-Amino-2-(4-fluorophenoxy)ethylidene]malononitrile

Hazard: Irritant

Formula: C₁₁H₈FN₃O
MW: 217.199 g/mol
Melting Pt: 189…191 °C
MDL Number: MFCD00139200
CAS Number: 1284228-23-4

Order Now

Specification Test Results

Melting point 189-191°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR