(1-Amino-2-phenoxyethylidene)malononitrile ≥95%

Supplier: Matrix Scientific

045218-500MG 045218-1G 045218-5G
101869-746EA 375.38 USD
101869-746 101869-748 101869-750
(1-Amino-2-phenoxyethylidene)malononitrile ≥95%
(1-Amino-2-phenoxyethylidene)malononitrile

Hazard: Irritant

Formula: C₁₁H₉N₃O
Melting Pt: 174…176 °C
CAS Number: 118645-79-7

Order Now

Specification Test Results

Melting point 174-176°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR