4'-(4-Methyl-1-piperazinyl)acetophenone ≥95%

Supplier: Matrix Scientific

Synonyms: 1-[4-(4-Methylpiperazino)phenyl]-1-ethanone

045217-500MG 045217-1G 045217-5G
101869-740EA 181.54 USD
101869-740 101869-742 101869-744
4'-(4-Methyl-1-piperazinyl)acetophenone ≥95%
4'-(4-Methyl-1-piperazinyl)acetophenone

Hazard: Irritant

Formula: C₁₃H₁₈N₂O
Melting Pt: 99-102°C
MDL Number: MFCD00023297
CAS Number: 26586-55-0

Order Now

Specification Test Results

Melting point 99-102°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR