[2-(4-Chlorophenoxy)-5-nitrophenyl]methanol ≥95%

Supplier: Matrix Scientific

045215-500MG 045215-1G 045215-5G
101869-728EA 375.38 USD
101869-728 101869-730 101869-732
[2-(4-Chlorophenoxy)-5-nitrophenyl]methanol ≥95%
[2-(4-Chlorophenoxy)-5-nitrophenyl]methanol

Hazard: Irritant

Formula: C₁₃H₁₀ClNO₄
Melting Pt: 118…120 °C
MDL Number: MFCD08056656
CAS Number: 886361-95-1

Order Now

Specification Test Results

Melting point 118-120°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR