2-(4-Chlorophenoxy)-5-nitrobenzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

045214-500MG 045214-1G 045214-5G
101869-722EA 278.46 USD
101869-722 101869-724 101869-726
2-(4-Chlorophenoxy)-5-nitrobenzenecarbaldehyde ≥95%
2-(4-Chlorophenoxy)-5-nitrobenzenecarbaldehyde

Hazard: Irritant

Formula: C₁₃H₈ClNO₄
Melting Pt: 115…116 °C
MDL Number: MFCD08056655
CAS Number: 708254-50-6

Order Now

Specification Test Results

Melting point 115-116°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR