2-Bromo-1-(1,3-thiazol-2-yl)-1-ethanonehydrobromide ≥95%

Supplier: Matrix Scientific

045212-500MG 045212-1G
101869-714EA 556.92 USD
101869-714 101869-716
2-Bromo-1-(1,3-thiazol-2-yl)-1-ethanonehydrobromide ≥95%
2-Bromo-1-(1,3-thiazol-2-yl)-1-ethanonehydrobromide

Hazard: Irritant

Formula: C₅H₅Br₂NOS
Melting Pt: 193…194 °C
MDL Number: MFCD08056654
CAS Number: 199804-81-4

Order Now

Specification Test Results

Melting point 193-194°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR