Ethyl-2-{2-[3-(trifluoromethyl)phenyl]-1,3-thiazol-4-yl}acetate ≥95%

Supplier: Matrix Scientific

045209-500MG 045209-1G
101869-702EA 375.38 USD
101869-702 101869-704
Ethyl-2-{2-[3-(trifluoromethyl)phenyl]-1,3-thiazol-4-yl}acetate ≥95%
Ethyl-2-{2-[3-(trifluoromethyl)phenyl]-1,3-thiazol-4-yl}acetate

Hazard: Irritant

Formula: C₁₄H₁₂F₃NO₂S
Melting Pt: 60…62 °C
MDL Number: MFCD08056653
CAS Number: 78743-00-7

Order Now

Specification Test Results

Melting point 60-62°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR