4-(1,3-Benzodioxol-5-yl)benzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

045208-500MG 045208-1G 045208-5G
101869-696EA 375.38 USD
101869-696 101869-698 101869-700
4-(1,3-Benzodioxol-5-yl)benzenecarbaldehyde ≥95%
4-(1,3-Benzodioxol-5-yl)benzenecarbaldehyde

Hazard: Irritant

Formula: C₁₄H₁₀O₃
MW: 226.23 g/mol
Melting Pt: 102…104 °C
MDL Number: MFCD07782410
CAS Number: 342889-39-8

Order Now

Specification Test Results

Melting point 102-104°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR