4-(3-Thienyl)benzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

045206-500MG 045206-1G 045206-5G
101869-684EA 346.15 USD
101869-684 101869-686 101869-688
4-(3-Thienyl)benzenecarbaldehyde ≥95%
4-(3-Thienyl)benzenecarbaldehyde

Hazard: Irritant

Formula: C₁₁H₈OS
Melting Pt: 98…100 °C
MDL Number: MFCD04039151
CAS Number: 157730-74-0

Order Now

Specification Test Results

Melting point 98-100°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR