2-(4-Bromophenyl)-5-nitrobenzoic acid ≥95%

Supplier: Matrix Scientific

045204-500MG 045204-1G 045204-5G
101869-672EA 747.69 USD
101869-672 101869-674 101869-676
2-(4-Bromophenyl)-5-nitrobenzoic acid ≥95%
2-(4-Bromophenyl)-5-nitrobenzoic acid

Hazard: Irritant

Formula: C₁₃H₈BrNO₄
Melting Pt: 221…223 °C
MDL Number: MFCD08056652
CAS Number: 886361-93-9

Order Now

Specification Test Results

Melting point 221-223°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR