Ethyl-3-trifluoromethyl-1H-pyrazole-5-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 3-(Trifluoromethyl)pyrazole-5-carboxylate

045201-500MG 045201-1G
101869-664EA 375.38 USD
101869-664 101869-666
Ethyl-3-trifluoromethyl-1H-pyrazole-5-carboxylate ≥97%
Ethyl-3-trifluoromethyl-1H-pyrazole-5-carboxylate

Hazard: Irritant

Formula: C₇H₇F₃N₂O₂
MW: 208.14 g/mol
Melting Pt: 95 °C
MDL Number: MFCD04111238
CAS Number: 129768-30-5

Order Now

Specification Test Results

Melting point 95°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR