2-Morpholinonicotinic acid ≥95%
Supplier: Matrix Scientific
045195-500MG
045195-1G
045195-5G
101869-628EA
181.54
USD
101869-628
101869-630
101869-632
2-Morpholinonicotinic acid ≥95%
2-Morpholinonicotinic acid
Hazard: Irritant
Formula:
C₁₀H₁₂N₂O₃ Melting Pt: 134…136 °C |
MDL Number:
MFCD03086193 CAS Number: 423768-54-1 |
Specification Test Results
Melting point | 134-136°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...