1-{3-Chloro-4-[(3-methoxybenzyl)oxy]phenyl}-1-ethanone ≥95%

Supplier: Matrix Scientific

045192-500MG 045192-1G 045192-5G
101869-612EA 375.38 USD
101869-612 101869-614 101869-616
1-{3-Chloro-4-[(3-methoxybenzyl)oxy]phenyl}-1-ethanone ≥95%
1-{3-Chloro-4-[(3-methoxybenzyl)oxy]phenyl}-1-ethanone

Hazard: Irritant

Formula: C₁₆H₁₅ClO₃
Melting Pt: 107…109 °C
MDL Number: MFCD06659577
CAS Number: 885949-84-8

Order Now

Specification Test Results

Melting point 107-109°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR