9H-Fluoren-9-ylmethyl-4-(4-hydroxyphenyl)tetrahydro-1(2H)-pyrazinecarboxylate ≥95%

Supplier: Matrix Scientific

045190-500MG 045190-1G 045190-5G
101869-600EA 278.46 USD
101869-600 101869-602 101869-604
9H-Fluoren-9-ylmethyl-4-(4-hydroxyphenyl)tetrahydro-1(2H)-pyrazinecarboxylate ≥95%
9H-Fluoren-9-ylmethyl-4-(4-hydroxyphenyl)tetrahydro-1(2H)-pyrazinecarboxylate

Hazard: Irritant

Formula: C₂₅H₂₄N₂O₃
MW: 400.48 g/mol
Melting Pt: 189…192 °C
MDL Number: MFCD06659575
CAS Number: 885949-83-7

Order Now

Specification Test Results

Melting point 189-192°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR