3-(4-Chlorophenyl)-3-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid ≥95%

Supplier: Matrix Scientific

045189-500MG 045189-1G 045189-5G
101869-594EA 375.38 USD
101869-594 101869-596 101869-598
3-(4-Chlorophenyl)-3-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid ≥95%
3-(4-Chlorophenyl)-3-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid

Hazard: Irritant

Formula: C₂₄H₂₀ClNO₄
Melting Pt: 193…195 °C
MDL Number: MFCD01863197
CAS Number: 194471-87-9

Order Now

Specification Test Results

Melting point 193-195°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR