1-{3-Fluoro-4-[(4-nitrobenzyl)oxy]phenyl}-1-ethanone ≥95%

Supplier: Matrix Scientific

045187-500MG 045187-1G 045187-5G
101869-582EA 375.38 USD
101869-582 101869-584 101869-586
1-{3-Fluoro-4-[(4-nitrobenzyl)oxy]phenyl}-1-ethanone ≥95%
1-{3-Fluoro-4-[(4-nitrobenzyl)oxy]phenyl}-1-ethanone

Hazard: Irritant

Formula: C₁₅H₁₂FNO₄
MW: 289.27 g/mol
Melting Pt: 148…150 °C
MDL Number: MFCD06659573
CAS Number: 885949-79-1

Order Now

Specification Test Results

Melting point 148-150°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR