3-Amino-3-(2,3-dichlorophenyl)propionic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Amino-3-(2,3-dichlorophenyl)propanoic acid

045185-500MG 045185-1G 045185-5G
101869-574EA 181.54 USD
101869-574 101869-576 101869-578
3-Amino-3-(2,3-dichlorophenyl)propionic acid ≥95%
3-Amino-3-(2,3-dichlorophenyl)propionic acid

Hazard: Irritant

Formula: C₉H₉Cl₂NO₂
MW: 234.0800 g/mol
Melting Pt: 232-235°C
MDL Number: MFCD01871301
CAS Number: 117391-56-7

Order Now

Specification Test Results

Melting point 232-235°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR