4-(3-Thienyl)-2-pyrimidinamine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Amino-4-(thien-3-yl)pyrimidine

045181-500MG 045181-1G 045181-5G
101869-552EA 375.38 USD
101869-552 101869-554 101869-556
4-(3-Thienyl)-2-pyrimidinamine ≥95%
4-(3-Thienyl)-2-pyrimidinamine

Hazard: Irritant

Formula: C₈H₇N₃S
MW: 177.23 g/mol
Melting Pt: 222…224 °C
MDL Number: MFCD06659572
CAS Number: 206564-03-6

Order Now

Specification Test Results

Melting point 222-224°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR