2-Amino-4-(4-bromophenyl)-5-pyrimidinecarbonitrile ≥95%

Supplier: Matrix Scientific

045179-500MG 045179-1G 045179-5G
101869-544EA 375.38 USD
101869-544 101869-546 101869-548
2-Amino-4-(4-bromophenyl)-5-pyrimidinecarbonitrile ≥95%
2-Amino-4-(4-bromophenyl)-5-pyrimidinecarbonitrile

Hazard: Irritant

Formula: C₁₁H₇N₄Br
Melting Pt: 258-260°C
MDL Number: MFCD06659571
CAS Number: 749885-93-6

Order Now

Specification Test Results

Melting point 258-260°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR