2-(4-Acetylphenoxy)-2-phenylacetic acid ≥95%
Supplier: Matrix Scientific
045175-500MG
045175-1G
045175-5G
101869-524EA
113.85
USD
101869-524
101869-526
101869-528
2-(4-Acetylphenoxy)-2-phenylacetic acid ≥95%
2-(4-Acetylphenoxy)-2-phenylacetic acid
Hazard: Irritant
Formula:
C₁₆H₁₄O₄ Melting Pt: 96-99°C |
CAS Number:
885949-44-0 |
Specification Test Results
Melting point | 96-99°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...