3-(1,3-Benzodioxol-5-yl)-5-methyl-1H-pyrazole ≥95%

Supplier: Matrix Scientific

045174-500MG 045174-1G 045174-5G
101869-518EA 307.69 USD
101869-518 101869-520 101869-522
3-(1,3-Benzodioxol-5-yl)-5-methyl-1H-pyrazole ≥95%
3-(1,3-Benzodioxol-5-yl)-5-methyl-1H-pyrazole

Hazard: Irritant

Formula: C₁₁H₁₀N₂O₂
Melting Pt: 141…143 °C
MDL Number: MFCD06411629
CAS Number: 937022-12-3

Order Now

Specification Test Results

Melting point 141-143°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR