5-(3-Thienyl)-1H-pyrazole ≥95%

Supplier: Matrix Scientific

045171-500MG 045171-1G 045171-5G
101869-500EA 249.23 USD
101869-500 101869-502 101869-504
5-(3-Thienyl)-1H-pyrazole ≥95%
5-(3-Thienyl)-1H-pyrazole

Hazard: Irritant

Formula: C₇H₆N₂S
MW: 150.2 g/mol
Melting Pt: 112…114 °C
MDL Number: MFCD00121014
CAS Number: 19933-25-6

Order Now

Specification Test Results

Melting point 112-114°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR