4-Bromo-N'-hydroxybenzenecarboximidamide ≥95%

Supplier: Matrix Scientific

045170-500MG 045170-1G 045170-5G
101869-494EA 181.54 USD
101869-494 101869-496 101869-498
4-Bromo-N'-hydroxybenzenecarboximidamide ≥95%
4-Bromo-N'-hydroxybenzenecarboximidamide

Hazard: Irritant

Formula: C₇H₇BrN₂O
Melting Pt: 142…144 °C
MDL Number: MFCD00019951
CAS Number: 69113-23-1

Order Now

Specification Test Results

Melting point 142-144°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR