2-(5-Formyl-2-methoxyphenoxy)-2-phenylacetic acid ≥95%

Supplier: Matrix Scientific

045169-500MG 045169-1G 045169-5G
101869-488EA 375.38 USD
101869-488 101869-490 101869-492
2-(5-Formyl-2-methoxyphenoxy)-2-phenylacetic acid ≥95%
2-(5-Formyl-2-methoxyphenoxy)-2-phenylacetic acid

Hazard: Irritant

Formula: C₁₆H₁₄O₅
Melting Pt: 159-161°C
CAS Number: 885949-43-9

Order Now

Specification Test Results

Melting point 159-161°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR