2-Cyclopropyl-5-hydroxy-1-benzofuran-3-carbaldehyde ≥95%

Supplier: Matrix Scientific

045168-500MG 045168-1G
101869-484EA 556.92 USD
101869-484 101869-486
2-Cyclopropyl-5-hydroxy-1-benzofuran-3-carbaldehyde ≥95%
2-Cyclopropyl-5-hydroxy-1-benzofuran-3-carbaldehyde

Hazard: Irritant

Formula: C₁₂H₁₀O₃
Melting Pt: 194…196 °C
MDL Number: MFCD11840949
CAS Number: 1135282-80-2

Order Now

Specification Test Results

Melting point 194-196°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR