1-(3,4-Dichlorophenyl)-2-[2-(methylsulfanyl)-4-pyrimidinyl]-1-ethanone ≥90%

Supplier: Matrix Scientific

045165-500MG 045165-1G 045165-5G
101869-466EA 556.92 USD
101869-466 101869-468 101869-470
1-(3,4-Dichlorophenyl)-2-[2-(methylsulfanyl)-4-pyrimidinyl]-1-ethanone ≥90%
1-(3,4-Dichlorophenyl)-2-[2-(methylsulfanyl)-4-pyrimidinyl]-1-ethanone

Hazard: Irritant

Formula: C₁₃H₁₀Cl₂N₂OS
Melting Pt: 135…136 °C
MDL Number: MFCD09027088
CAS Number: 937604-22-3

Order Now

Specification Test Results

Melting point 135-136°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR