1-Boc-4-(phenylamino)piperidine ≥95%
Supplier: Matrix Scientific
Synonyms:
4-(Phenylamino)piperidine, N1-BOC protected
045161-500MG
045161-1G
045161-5G
101869-442EA
278.46
USD
101869-442
101869-444
101869-446
1-Boc-4-(phenylamino)piperidine ≥95%
1-Boc-4-(phenylamino)piperidine
Hazard: Irritant
Formula:
C₁₆H₂₄N₂O₂ Melting Pt: 136…137 °C |
CAS Number:
125541-22-2 |
Specification Test Results
Melting point | 136-137°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...