1-[1-(4-Bromophenyl)-2,5-dimethyl-1H-pyrrol-3-yl]-2,2,2-trifluoro-1-ethanone ≥95%

Supplier: Matrix Scientific

045157-500MG 045157-1G 045157-5G
101869-418EA 278.46 USD
101869-418 101869-420 101869-422
1-[1-(4-Bromophenyl)-2,5-dimethyl-1H-pyrrol-3-yl]-2,2,2-trifluoro-1-ethanone ≥95%
1-[1-(4-Bromophenyl)-2,5-dimethyl-1H-pyrrol-3-yl]-2,2,2-trifluoro-1-ethanone

Hazard: Irritant

Formula: C₁₄H₁₁BrF₃NO
Melting Pt: 79…82 °C
MDL Number: MFCD00268899
CAS Number: 95337-69-2

Order Now

Specification Test Results

Melting point 79-82°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR